Merck
Atomic scale modeling of iron-doped biphasic calcium phosphate bioceramics.
Acta biomaterialia (2016-12-15)
Sandrine Gomes, Amandeep Kaur, Jean-Marc Grenèche, Jean-Marie Nedelec, Guillaume Renaudin
ABSTRAKT
Biphasic calcium phosphates (BCPs) are bioceramics composed of hydroxyapatite (HAp, Ca Biphasic calcium phosphates (BCPs) are bioceramics composed of hydroxyapatite (HAp, Ca