Merck
5-(2-Pyridil)-1H-tetrazole complexes with Mo(iv) and W(iv) cyanides.
Dalton transactions (Cambridge, England : 2003) (2017-03-09)
A Mrowiec, A Jurowska, M Hodorowicz, J Szklarzewicz
ABSTRAKT
The reactions of tetrazole ligand derived from 2-cyanopyridine with K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%