Merck
Rendering High Surface Area, Mesoporous Metal-Organic Frameworks Electronically Conductive.
ACS applied materials & interfaces (2017-03-21)
Timothy C Wang, Idan Hod, Cornelius O Audu, Nicolaas A Vermeulen, SonBinh T Nguyen, Omar K Farha, Joseph T Hupp
ABSTRAKT
We report the design and synthesis of a metal-organic framework (MOF)-polythiophene composite that has comparable electronic conductivity to reported conductive 3-D MOFs, but with display and retention of high porosity, including mesoporosity. A robust zirconium MOF, NU-1000, was rendered electronically conductive by first incorporating, via solvent-assisted ligand incorporation (SALI), a carefully designed pentathiophene derivative at a density of one pentamer per hexa-zirconium node. Using a cast film of the intermediate composite (termed pentaSALI) on conductive glass, the incorporated oligothiophene was electrochemically polymerized to yield the conductive composite, Epoly. Depending on the doping level of the polythiophene in the composite, it can be tuned from an insulating state to a semiconduting state with conductivity of 1.3 × 10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,2′-Bithiophene, 99%
Sigma-Aldrich
3-Bromothiophene, 97%