Merck
Zebrafish have an ethanol-inducible hepatic 4-nitrophenol hydroxylase that is not CYP2E1-like.
Environmental toxicology and pharmacology (2017-07-21)
Jessica H Hartman, Jordan S Kozal, Richard T Di Giulio, Joel N Meyer
ABSTRAKT
Zebrafish are an attractive model organism for toxicology; however, an important consideration in translating between species is xenobiotic metabolism/bioactivation. CYP2E1 metabolizes small hydrophobic molecules, e.g. ethanol, cigarette smoke, and diesel exhaust components. CYP2E1 is thought to only be conserved in mammals, but recent reports identified homologous zebrafish cytochrome P450s. Herein, ex vivo biochemical measurements show that unlike mammals, zebrafish possess a low-affinity 4-nitrophenol hydroxylase (K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fomepizole, 99%