Merck
Maternal metabolic stress may affect oviduct gatekeeper function.
Reproduction (Cambridge, England) (2017-03-05)
L Jordaens, V Van Hoeck, V Maillo, A Gutierrez-Adan, W F A Marei, B Vlaeminck, S Thys, R G Sturmey, P E J Bols, J L M R Leroy
ABSTRAKT
We hypothesized that elevated non-esterified fatty acids (NEFA) modify
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyridinium chlorochromate, 98%