Merck
  • Development of a long-term serum-free culture system for immature granulosa cells from diethylstilboestrol-treated prepubertal rabbits: influence of androstenedione and fibronectin on FSH-induced cytodifferentiation
Development of a long-term serum-free culture system for immature granulosa cells from diethylstilboestrol-treated prepubertal rabbits: influence of androstenedione and fibronectin on FSH-induced cytodifferentiation
Reproduction (Cambridge, England) (2000)
Picazo, R., et al.