Merck
The antiviral drug ribavirin is a selective inhibitor of S-adenosyl-L-homocysteine hydrolase from Trypanosoma cruzi.
Bioorganic & medicinal chemistry (2007-09-12)
Sumin Cai, Qing-Shan Li, Ronald T Borchardt, Krzysztof Kuczera, Richard L Schowen
ABSTRAKT
Ribavirin (1,2,4-triazole-3-carboxamide riboside) is a well-known antiviral drug. Ribavirin has also been reported to inhibit human S-adenosyl-L-homocysteine hydrolase (Hs-SAHH), which catalyzes the conversion of S-adenosyl-L-homocysteine to adenosine and homocysteine. We now report that ribavirin, which is structurally similar to adenosine, produces time-dependent inactivation of Hs-SAHH and Trypanosoma cruzi SAHH (Tc-SAHH). Ribavirin binds to the adenosine-binding site of the two SAHHs and reduces the NAD(+) cofactor to NADH. The reversible binding step of ribavirin to Hs-SAHH and Tc-SAHH has similar K(I) values (266 and 194 microM), but the slow inactivation step is 5-fold faster with Tc-SAHH. Ribavirin may provide a structural lead for design of more selective inhibitors of Tc-SAHH as potential anti-parasitic drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ribavirin, antiviral