Merck
A new cycloartane triterpenoid glycoside from Souliea vaginata.
Natural product research (2017-04-14)
Hai-Feng Wu, Xin Liu, Yin-Di Zhu, Jing Zhou, Ying-Ying Gong, Guo-Xu Ma, Xu-Dong Xu, Yi-Lin Liu, Zheng-Hong Luo, Di-Zhao Chen, Qiong-Yu Zou, Zi-Jian Zhao
ABSTRAKT
One new cycloartane triterpenoid glycoside, soulieoside Q (1), together with four known compounds (2-5) were isolated from the ethanolic extract of the rhizomes of Souliea vaginata Maxim. The structure of the new compound was determined by extensive spectroscopic analysis including 1D and 2D NMR and HRESIMS, as well as chemical methods. Compound 1 was evaluated for its cytotoxic activities against HepG2 and A549 cancer cell lines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%