Merck
A rapid administration of GW4064 inhibits the NLRP3 inflammasome activation independent of farnesoid X receptor agonism.
FEBS letters (2017-08-09)
Shujun Xie, Chuansheng Guo, Zhexu Chi, Bo Huang, Yihua Wu, Di Wang, Dajing Xia
ABSTRAKT
GW4064 is a small molecule known to be an agonist of the nuclear farnesoid X receptor (FXR). We found that GW4064 inhibits the NLR family CARD domain containing 3 (NLRP3) inflammasome activation in an FXR-independent manner as evidenced by its similar inhibitory effect on NLRP3 inflammasome activation in FXR-deficient macrophages. Interestingly, GW4064 decreases the nigericin-induced oligomerization and ubiquitination of ASC which is critical for the NLRP3 inflammasome activation. In vivo results indicate that GW4064 could partially rescue the symptoms of NLRP3-dependent inflammatory disease models. These results not only necessitate cautious interpretation of the biological function of GW4064 as an FXR agonist, but also provide a potential therapeutic approach using GW4064 in the treatment of NLRP3-related diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder