Merck
β
Pharmacological research (2017-08-02)
Emma Mitidieri, Teresa Tramontano, Danila Gurgone, Ciro Imbimbo, Vincenzo Mirone, Ferdinando Fusco, Giuseppe Cirino, Roberta d'Emmanuele di Villa Bianca, Raffaella Sorrentino
ABSTRAKT
Erectile function is a widely accepted indicator of systemic endothelial activity since from a clinical standpoint erectile dysfunction (ED) often precedes cardiovascular events. Recently it has been described a potential role for β
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor