Merck
pH-Responsive quencher-free molecular beacon systems containing 2'-deoxyuridine units labeled with fluorene derivatives.
Organic & biomolecular chemistry (2017-08-16)
Ji Won Lee, Ye-Seul Son, Jun Yeon Hwang, Yoojin Park, Gil Tae Hwang
ABSTRAKT
Three fluorescent nucleosides-U
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)