Merck
Experimental and therapeutic medicine (2017-10-19)
Liangfeng Tang, Haijun Zhong, Hong Chen, Jian Shen, Yunli Bi, Xianmin Xiao
ABSTRAKT
Fetal repair of myelomeningocele (MMC) has been proven to be beneficial for the central nervous system development; however, the effect of fetal MMC repair on bladder function remains controversial. The objective of the present study was to establish an early timepoint for
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder