Merck
EGF hijacks miR-198/FSTL1 wound-healing switch and steers a two-pronged pathway toward metastasis.
The Journal of experimental medicine (2017-08-23)
Gopinath M Sundaram, Hisyam M Ismail, Mohsin Bashir, Manish Muhuri, Candida Vaz, Srikanth Nama, Ghim Siong Ow, Ivshina Anna Vladimirovna, Rajkumar Ramalingam, Brian Burke, Vivek Tanavde, Vladimir Kuznetsov, E Birgitte Lane, Prabha Sampath
ABSTRAKT
Epithelial carcinomas are well known to activate a prolonged wound-healing program that promotes malignant transformation. Wound closure requires the activation of keratinocyte migration via a dual-state molecular switch. This switch involves production of either the anti-migratory microRNA miR-198 or the pro-migratory follistatin-like 1 (FSTL1) protein from a single transcript; miR-198 expression in healthy skin is down-regulated in favor of FSTL1 upon wounding, which enhances keratinocyte migration and promotes re-epithelialization. Here, we reveal a defective molecular switch in head and neck squamous cell carcinoma (HNSCC). This defect shuts off miR-198 expression in favor of sustained FSTL1 translation, driving metastasis through dual parallel pathways involving DIAPH1 and FSTL1. DIAPH1, a miR-198 target, enhances directional migration through sequestration of Arpin, a competitive inhibitor of Arp2/3 complex. FSTL1 blocks Wnt7a-mediated repression of extracellular signal-regulated kinase phosphorylation, enabling production of MMP9, which degrades the extracellular matrix and facilitates metastasis. The prognostic significance of the FSTL1-DIAPH1 gene pair makes it an attractive target for therapeutic intervention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Anti-Arpin Antibody, from rabbit, purified by affinity chromatography