- The pan-BCL-2-blocker obatoclax (GX15-070) and the PI3-kinase/mTOR-inhibitor BEZ235 produce cooperative growth-inhibitory effects in ALL cells.
The pan-BCL-2-blocker obatoclax (GX15-070) and the PI3-kinase/mTOR-inhibitor BEZ235 produce cooperative growth-inhibitory effects in ALL cells.
Oncotarget (2017-10-06)
Gabriele Stefanzl, Daniela Berger, Sabine Cerny-Reiterer, Katharina Blatt, Gregor Eisenwort, Wolfgang R Sperr, Gregor Hoermann, Karin Lind, Alexander W Hauswirth, Peter Bettelheim, Heinz Sill, Junia V Melo, Ulrich Jäger, Peter Valent
PMID28978065
ABSTRAKT
Acute lymphoblastic leukemia (ALL) is characterized by leukemic expansion of lymphoid blasts in hematopoietic tissues. Despite improved therapy only a subset of patients can be cured. Therefore, current research is focusing on new drug-targets. Members of the BCL-2 family and components of the PI3-kinase/mTOR pathway are critically involved in the regulation of growth and survival of ALL cells. We examined the effects of the pan-BCL-2 blocker obatoclax and the PI3-kinase/mTOR-inhibitor BEZ235 on growth and survival of ALL cells. In