Merck
Two-Photon Imaging of 3D Organization of Bimetallic AuAg Nanoclusters in DNA Matrix.
Langmuir : the ACS journal of surfaces and colloids (2017-08-13)
Katarzyna Brach, Magdalena Waszkielewicz, Joanna Olesiak-Banska, Marek Samoc, Katarzyna Matczyszyn
ABSTRAKT
We report on two-photon excitation properties of small silver-doped gold nanoclusters (AuAgNCs) and on their three-dimensional arrangement in a hybrid system composed of DNA liquid crystals (LCs) and AuAgNCs. UV-vis and fluorescence spectroscopy, transmission electron microscopy (TEM), and multiphoton excitation spectroscopy were used to characterize the nanoparticles. We show that AuAgNCs exhibit two-photon excited luminescence (2PL) emission and second-harmonic generation (SHG) and that these properties remain the same in liquid crystalline matrix. The results are described in detail and discussed in the context of possible imaging application of AuAgNC and specific AuAgNCs organization induced by liquid crystalline ordering of DNA molecules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %