Merck
  • E2F site in the essential promoter region does not confer S phase-specific transcription of the ABCC10 gene in human prostate cancer cells.
E2F site in the essential promoter region does not confer S phase-specific transcription of the ABCC10 gene in human prostate cancer cells.
Acta biochimica Polonica (2017-06-15)
Magdalena Dabrowska, Francis M Sirotnak
ABSTRAKT
ABCC10 (MRP7) plays a role in cellular detoxification and resistance to anticancer drugs. Since ABCC10 gene transcription in human prostate cancer CWR22Rv1 cells was found dependent on E2F binding sequence motif, ABCC10 expression in G
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Mimosine from Koa hoale seeds, ≥98%