- Tungsten accumulation by highly tolerant marine hydrothermal Sulfitobacter dubius strains carrying a tupBCA cluster.
Tungsten accumulation by highly tolerant marine hydrothermal Sulfitobacter dubius strains carrying a tupBCA cluster.
Systematic and applied microbiology (2017-08-15)
Carina Coimbra, Pedro Farias, Rita Branco, Paula V Morais
PMID28802743
ABSTRAKT
Tungsten (W) has industrial and economic importance, and is in the European Union list of metals with a high supply risk. It is used by living organisms, which transport it into the cell, in the form of tungstate ion (WO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Tungsten, tube, outside diameter 1.6 mm, length 50 mm, wall thickness 0.5 mm