Merck
Integrating a Semitransparent, Fullerene-Free Organic Solar Cell in Tandem with a BiVO
ACS applied materials & interfaces (2017-06-22)
Yuelin Peng, Gokul V Govindaraju, Dong Ki Lee, Kyoung-Shin Choi, Trisha L Andrew
ABSTRAKT
We report an unassisted solar water splitting system powered by a diketopyrrolopyrrole (DPP)-containing semitransparent organic solar cell. Two major merits of this fullerene-free solar cell enable its integration with a BiVO