Merck
A class-selective immunoassay for simultaneous analysis of anilinopyrimidine fungicides using a rationally designed hapten.
The Analyst (2017-09-29)
F A Esteve-Turrillas, J V Mercader, C Agulló, A Abad-Somovilla, A Abad-Fuentes
ABSTRAKT
The development of multianalyte immunoassays constitutes a main research issue in the field of bioanalytical techniques. In the present study, class-specific antibodies against the three members of the anilinopyrimidine family of fungicides (pyrimethanil, cyprodinil and mepanipyrim) were raised by using a bioconjugate of a rationally designed hapten [5-(6-methyl-2-(phenylamino)pyrimidin-4-yl)pentanoic acid]. Highly sensitive immunoassays were developed for the generic determination of these compounds, using the competitive enzyme-linked immunosorbent assay (ELISA). Particularly, a direct antibody-coated competitive ELISA afforded identical sensitivity for the three anilinopyrimidines, with IC
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-(N-Anilino)pyrimidine
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)