Merck
CaMKII-mediated Beclin 1 phosphorylation regulates autophagy that promotes degradation of Id and neuroblastoma cell differentiation.
Nature communications (2017-10-29)
Xuan Li, Xiao-Qi Wu, Rong Deng, Dan-Dan Li, Jun Tang, Wen-Dan Chen, Jing-Hong Chen, Jiao Ji, Lin Jiao, Shan Jiang, Fen Yang, Gong-Kan Feng, Ravichandran Senthilkumar, Fei Yue, Hai-Liang Zhang, Rui-Yan Wu, Yan Yu, Xue-Lian Xu, Jia Mai, Zhi-Ling Li, Xiao-Dan Peng, Yun Huang, Xiang Huang, Ning-Fang Ma, Qian Tao, Yi-Xin Zeng, Xiao-Feng Zhu
ABSTRAKT
Autophagy is a degradative pathway that delivers cellular components to the lysosome for degradation. The role of autophagy in cell differentiation is poorly understood. Here we show that CaMKII can directly phosphorylate Beclin 1 at Ser90 to promote K63-linked ubiquitination of Beclin 1 and activation of autophagy. Meanwhile, CaMKII can also promote K63-linked ubiquitination of inhibitor of differentiation 1/2 (Id-1/2) by catalyzing phosphorylation of Id proteins and recruiting TRAF-6. Ubiquitinated Id-1/Id-2 can then bind to p62 and be transported to autolysosomes for degradation. Id degradation promotes the differentiation of neuroblastoma cells and reduces the proportion of stem-like cells. Our study proposes a mechanism by which autophagic degradation of Id proteins can regulate cell differentiation. This suggests that targeting of CaMKII and the regulation of autophagic degradation of Id may be an effective therapeutic strategy to induce cell differentiation in neuroblastoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)