Merck
  • Use of Plant Preservative Mixture™ for establishing in vitro cultures from field plants: Experience with papaya reveals several PPM™ tolerant endophytic bacteria.
Use of Plant Preservative Mixture™ for establishing in vitro cultures from field plants: Experience with papaya reveals several PPM™ tolerant endophytic bacteria.
Plant cell reports (2017-07-28)
Pious Thomas, Mukta Agrawal, C B Bharathkumar
ABSTRAKT
Prevalence of diverse PPM™-tolerant endophytic bacteria in papaya, the broad-spectrum microbicide specified for use in plant tissue cultures, capable of surviving covertly in MS-based medium, with implications in contamination management. Plant Preservative Mixture™ was employed for establishing papaya (Carica papaya) tissue cultures from field explants. Comparing three recommended practices for controlling endogenous microbial contaminants, axillary shoot tips (1.0-1.5 cm) from cv. Arka Prabhath were treated with PPM™ 5% for 4 h (T1), 50% for 10 min (T2) or 100% for 10 min (T3) and cultured in MS-based papaya establishment medium (PEM). By 4-6 weeks, all treatments proved non-rewarding with cultures succumbing either to microbial contamination (80% in T1) or phytotoxicity effect/contamination (90% in T2 and 95% in T3). Another trial adopting a multi-step surface sterilization treatment (carbendazim-cetrimide-HgCl