Merck
Pinus pinaster Knot: A Source of Polyphenols against Plasmopara viticola.
Journal of agricultural and food chemistry (2017-09-20)
Julien Gabaston, Tristan Richard, Stéphanie Cluzet, Antonio Palos Pinto, Marie-Cécile Dufour, Marie-France Corio-Costet, Jean-Michel Mérillon
ABSTRAKT
Pine knot extract from Pinus pinaster byproducts was characterized by UHPLC-DAD-MS and NMR. Fourteen polyphenols divided into four classes were identified as follows: lignans (nortrachelogenin, pinoresinol, matairesinol, isolariciresinol, secoisolariciresinol), flavonoids (pinocembrin, pinobanksin, dihydrokaempferol, taxifolin), stilbenes (pinosylvin, pinosylvin monomethyl ether, pterostilbene), and phenolic acids (caffeic acid, ferulic acid). The antifungal potential of pine knot extract, as well as the main compounds, was tested in vitro against Plasmopara viticola. The ethanolic extract showed a strong antimildew activity. In addition, pinosylvins and pinocembrin demonstrated significant inhibition of zoospore mobility and mildew development. These findings strongly suggest that pine knot is a potential biomass that could be used as a natural antifungal product.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pterostilbene, ≥97% (HPLC), solid