Merck
Correlation between oral fluid and plasma oxytetracycline concentrations after intramuscular administration in pigs.
Journal of veterinary pharmacology and therapeutics (2017-05-01)
A Gajda, A Jablonski, M Gbylik-Sikorska, A Posyniak
ABSTRAKT
The penetration of oxytetracycline (OTC) into the oral fluid and plasma of pigs and correlation between oral fluid and plasma were evaluated after a single intramuscular (i.m.) dose of 20 mg/kg body weight of long-acting formulation. The OTC was detectable both in oral fluid and plasma from 1 hr up to 21 day after drug administration. The maximum concentrations (C