Merck
Fluorogenic mercury ion sensor based on pyrene-amino mercapto thiadiazole unit.
Journal of hazardous materials (2017-09-25)
B Kirthika Rani, S Abraham John
ABSTRAKT
A highly selective and sensitive determination of Hg
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Pyrenecarboxaldehyde, 99%