Merck
Degradation of Acid Orange 7 using peroxymonosulfate catalyzed by granulated activated carbon and enhanced by electrolysis.
Chemosphere (2017-09-08)
Jing Li, Heng Lin, Kangmeng Zhu, Hui Zhang
ABSTRAKT
Electrochemistry coupled with granulated activated carbon catalysis of peroxymonosulfate (electro/GAC/PMS) as a novel wastewater treatment process was performed for the degradation of Acid Orange 7 (AO7) in aqueous solution. The decolorization of AO7 was compared under different permutations and combinations of electro-oxidation, GAC and PMS. It showed that the electro/GAC/PMS process was the most effective and the decolorization of AO7 followed pseudo-first order kinetics. The surface chemistry of GAC samples was analyzed by Fourier transform infrared spectroscopy and X-ray photoelectron spectroscopy. Compared with the fresh samples, π-electron density and hydroxyl group content decreased under the GAC/PMS system, but kept the similar values under the electro/GAC/PMS system. Electron paramagnetic resonance and radical scavenger studies were used to verify the formation of sulfate radicals (SO