Merck
Fully disposable microfluidic electrochemical device for detection of estrogen receptor alpha breast cancer biomarker.
Biosensors & bioelectronics (2017-07-30)
Carolina V Uliana, Camila R Peverari, André S Afonso, Marcia R Cominetti, Ronaldo C Faria
ABSTRAKT
A novel fully disposable microfluidic electrochemical array device (µFED) was developed and successfully applied for detection of the biomarker estrogen receptor alpha (ERα). The µFED was constructed using low-cost materials and an inexpensive home cutter printer enabled the manufacture of dozens of µFEDs in less than 2h, at a cost of less than US$ 0.20 in material per device. The µFED incorporates counter and reference electrodes and eight carbon-based working electrodes, which were modified with DNA sequences known as estrogen response elements (DNA-ERE), where ERα binds specifically. Paramagnetic particles heavily decorated with anti-ERα antibody and horseradish peroxidase (MP-Ab-HRP) were used to efficiently capture ERα from the sample solution. The ERα-MP-Ab-HRP bioconjugate formed was injected into the µFED and incubated with the DNA-ERE-modified electrodes, followed by amperometric detection with application of -0.2V vs. Ag|AgCl while a mixture of H
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%