Merck
  • Fibroblast growth factor 2 protects against renal ischaemia/reperfusion injury by attenuating mitochondrial damage and proinflammatory signalling.
Fibroblast growth factor 2 protects against renal ischaemia/reperfusion injury by attenuating mitochondrial damage and proinflammatory signalling.
Journal of cellular and molecular medicine (2017-05-26)
Xiao-Hua Tan, Xiao-Meng Zheng, Li-Xia Yu, Jian He, Hong-Mei Zhu, Xiu-Ping Ge, Xiao-Li Ren, Fa-Qing Ye, Saverio Bellusci, Jian Xiao, Xiao-Kun Li, Jin-San Zhang
ABSTRAKT
Ischaemia-reperfusion injury (I/RI) is a common cause of acute kidney injury (AKI). The molecular basis underlying I/RI-induced renal pathogenesis and measures to prevent or reverse this pathologic process remains to be resolved. Basic fibroblast growth factor (FGF2) is reported to have protective roles of myocardial infarction as well as in several other I/R related disorders. Herein we present evidence that FGF2 exhibits robust protective effect against renal histological and functional damages in a rat I/RI model. FGF2 treatment greatly alleviated I/R-induced acute renal dysfunction and largely blunted I/R-induced elevation in serum creatinine and blood urea nitrogen, and also the number of TUNEL-positive tubular cells in the kidney. Mechanistically, FGF2 substantially ameliorated renal I/RI by mitigating several mitochondria damaging parameters including pro-apoptotic alteration of Bcl2/Bax expression, caspase-3 activation, loss of mitochondrial membrane potential and K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
JC-1, powder or solid (Crystals)