Merck
Novel combinations of PI3K-mTOR inhibitors with dacomitinib or chemotherapy in PTEN-deficient patient-derived tumor xenografts.
Oncotarget (2017-11-22)
Irene Brana, Nhu-An Pham, Lucia Kim, Shingo Sakashita, Ming Li, Christine Ng, Yuhui Wang, Peter Loparco, Rafael Sierra, Lisa Wang, Blaise A Clarke, Benjamin G Neel, Lillian L Siu, Ming-Sound Tsao
ABSTRAKT
PTEN inactivation occurs commonly in human cancers and putatively activates the PI3K/AKT/ mTOR pathway. Activation of this pathway has been involved in resistance to chemotherapy or anti-EGFR/HER2 therapies. We evaluated the combination of PI3K-mTOR inhibitors with chemotherapy or the pan-HER inhibitor dacomitinib in PTEN-deficient patient-derived tumor xenografts (PDX). Three PDXs were selected for their lack of PTEN expression by immunohistochemistry: a triple-negative breast cancer (TNBC), a