Merck
Therapeutic potential of the phosphino Cu(I) complex (HydroCuP) in the treatment of solid tumors.
Scientific reports (2017-10-27)
Valentina Gandin, Cecilia Ceresa, Giovanni Esposito, Stefano Indraccolo, Marina Porchia, Francesco Tisato, Carlo Santini, Maura Pellei, Cristina Marzano
ABSTRAKT
[Cu(thp)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade