Merck
Aminoquinoline Fluorescent Labels Obstruct Efficient Removal of N-Glycan Core α(1-6) Fucose by Bovine Kidney α-l-Fucosidase (BKF).
Journal of proteome research (2017-09-28)
Róisín O'Flaherty, Aoife M Harbison, Philip J Hanley, Christopher H Taron, Elisa Fadda, Pauline M Rudd
ABSTRAKT
Here we report evidence that new aminoquinoline N-glycan fluorescent labels interfere with the release of core α(1-6) fucose from N-glycans by bovine kidney α-l-fucosidase (BKF). BKF is a commonly employed exoglycosidase for core α(1-6) fucose determination. Molecular simulations of the bound and unbound Fuc-α(1-6)-GlcNAc, where GlcNAc is situated at the reducing end for all N-glycans, suggest that the reduced BKF activity may be due to a nonoptimal fit of the highest populated conformers in the BKF active binding site at room temperature. Population analysis and free energy estimates suggest that an enhanced flexibility of the labeled sugar, which facilitates recognition and binding, can be achievable with extended reaction conditions. We provide these experimental conditions using a sequential exoglycosidase digestion array using high concentrations of BKF.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
6, 2′, 4′-trimethoxyflavone, ≥98% (HPLC)