Merck
Synthesis of kinase inhibitors containing a pentafluorosulfanyl moiety.
Organic & biomolecular chemistry (2017-10-07)
Supojjanee Sansook, Cory A Ocasio, Iain J Day, Graham J Tizzard, Simon J Coles, Oleg Fedorov, James M Bennett, Jonathan M Elkins, John Spencer
ABSTRAKT
A series of 3-methylidene-1H-indol-2(3H)-ones substituted with a 5- or 6-pentafluorosulfanyl group has been synthesized by a Knoevenagel condensation reaction of SF
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrrole-2-carboxaldehyde, 98%