- Magnetic dispersive solid-phase extraction based on modified magnetic nanoparticles for the detection of cocaine and cocaine metabolites in human urine by high-performance liquid chromatography-mass spectrometry.
Magnetic dispersive solid-phase extraction based on modified magnetic nanoparticles for the detection of cocaine and cocaine metabolites in human urine by high-performance liquid chromatography-mass spectrometry.
Journal of separation science (2017-08-27)
Feiyu Yang, Yun Zou, Chunfang Ni, Rong Wang, Min Wu, Chen Liang, Jiabin Zhang, Xiaoliang Yuan, Wenbin Liu
PMID28843028
ABSTRAKT
An easy-to-handle magnetic dispersive solid-phase extraction procedure was developed for preconcentration and extraction of cocaine and cocaine metabolites in human urine. Divinyl benzene and vinyl pyrrolidone functionalized silanized Fe