Merck
Emodin Alleviates Sodium Taurocholate-Induced Pancreatic Acinar Cell Injury
Frontiers in immunology (2017-11-23)
Hong Xiang, Xufeng Tao, Shilin Xia, Jialin Qu, Huiyi Song, Jianjun Liu, Dong Shang
ABSTRAKT
Pancreatitis is an inflammatory disease that is responsible for substantial morbidity and mortality, and it can induce pancreatic necrosis that starts within pancreatic acinar cells in severe cases. Emodin, a pleiotropic natural product isolated from the Chinese herb
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard