Merck
Paracrine interactions between mesenchymal stem cells and macrophages are regulated by 1,25-dihydroxyvitamin D3.
Scientific reports (2017-11-04)
Laura Saldaña, Gema Vallés, Fátima Bensiamar, Francisco José Mancebo, Eduardo García-Rey, Nuria Vilaboa
ABSTRAKT
Mesenchymal stem cells (MSC) modulate the macrophage-mediated inflammatory response through the secretion of soluble factors. In addition to its classical effects on calcium homeostasis, 1,25-dihydroxyvitamin D3 (1,25D3) has emerged as an important regulator of the immune system. The present study investigates whether 1,25D3 modulates the paracrine interactions between MSC and macrophages. 1,25D3 stimulated MSC to produce PGE