Merck
Ultradeformable liposome loaded with zinc phthalocyanine and [Ru(NH.NHq)(tpy)NO]
Photodiagnosis and photodynamic therapy (2017-06-05)
Renata Galvão de Lima, Antonio Cláudio Tedesco, Roberto Santana da Silva, Margaret Jayne Lawrence
ABSTRAKT
Ultradeformable liposomes (UDLs) as a drug delivery system (DDS), prepared from the unsaturated phospholipid, dioleylphosphocholine (DOPC), and containing the non-ionic surfactant Tween 20 as edge activator, have been explored as topical vehicles for zinc phthalocyanine (ZnPc) and the nitrosyl ruthenium complex [Ru(NH.NHq)(tpy)NO]