Merck
The prevalence of PI*S and PI*Z SERPINA1 alleles in healthy individuals and COPD patients in Saudi Arabia: A case-control study.
Medicine (2017-10-20)
Noura Al-Jameil, Amina A Hassan, Rana Hassanato, Sree R Isac, Maram Al Otaiby, Fadwa Al-Shareef, Basmah Al-Maarik, Iman Al Ajeyan, Khloud Al-Bahloul, Samina Ghani, Dana Al-Torbak
ABSTRAKT
Alpha-1 antitrypsin (AAT) is an acute phase protein produced in hepatocytes. Its deficiency affects the lungs and liver. A case-control study was carried out to determine the prevalence of 2 common deficiency alleles, PI*S and PI*Z, for alpha-1 antitrypsin deficiency (AATD) in both healthy and chronic obstructive pulmmonary disease (COPD)-affected Saudi populations and to clarify the importance of genetic tests in the screening of people at risk for COPD.One thousand blood samples from healthy individuals and 1000 from COPD-affected Saudi individuals were genotyped for the above-mentioned alleles, using real-time polymerase chain reaction (PCR), with the exclusion of any other nationalities. Data were analyzed by determining the allele and genotype frequencies through gene counting and its confidence intervals. The allele frequencies, derived by the Hardy-Weinberg equilibrium method, were analyzed by Pearson Chi-squared tests. The confidence intervals for genotype frequencies were calculated using exploratory software for confidence intervals.Of the 1000 COPD patients included in our study, the prevalence of PI*S and PI*Z was 21.8% and 7.7%, respectively, while within the 1000 normal samples, these alleles occurred in 8.9% of patients for PI*S and 1.6% for PI*Z. The AAT deficiency genotype frequencies (PI*ZZ, PI*SS, and PI*SZ) were 6.5 per 1000 and 87 per 1000 for normal and COPD-affected Saudi individuals.Our results indicated a high prevalence of AATD alleles in the normal Saudi population and an association between AAT deficiency and pulmonary disease development. Additionally, our research confirms the importance of genetic screening to achieve early and accurate diagnosis of AATD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %