Merck
Evaluation of a Picture-Based Test for the Assessment of Gelotophobia.
Frontiers in psychology (2017-12-07)
Willibald Ruch, Tracey Platt, Richard Bruntsch, Róbert Ďurka
ABSTRAKT
This study examines whether coding open answers in a picture-based test, as to the extent they reflect the fear of being laughed at (i.e., gelotophobia), demonstrates sufficient validity to construct a semi-projective test for the assessment of gelotophobia. Previous findings indicate that cartoon stimuli depicting laughter situations (i.e., in the pilot version of the Picture-Geloph; Ruch et al., 2009) on average elicit fear-typical responses in gelotophobes stronger than in non-gelotophobes. The present study aims to (a) develop a standardized scoring procedure based on a coding scheme, and (b) examine the properties of the pilot version of the Picture-Geloph in order to select the most acceptable items for a standard form of the test. For Study 1, a sample of
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lysergic acid diethylamide