Merck
Amplification of arsenic genotoxicity by TiO
Nanotoxicology (2017-10-20)
Xinan Wang, Yun Liu, Juan Wang, Yaguang Nie, Shaopeng Chen, Tom K Hei, Zhaoxiang Deng, Lijun Wu, Guoping Zhao, An Xu
ABSTRAKT
Titanium dioxide nanoparticles (TiO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
1-Adamantylamine, 97%
Sigma-Aldrich
Nystatin, ≥4,400 units/mg