- Glucagon-like peptide-1 potentiates glucose-stimulated insulin secretion via the transient receptor potential melastatin 2 channel.
Glucagon-like peptide-1 potentiates glucose-stimulated insulin secretion via the transient receptor potential melastatin 2 channel.
Experimental and therapeutic medicine (2017-12-05)
Bo Pang, Sungjoon Kim, Daiqing Li, Zejun Ma, Bei Sun, Xiaona Zhang, Zhongming Wu, Liming Chen
PMID29201240
ABSTRAKT
The transient receptor potential melastatin 2 (TRPM2) channel, a Ca
MATERIAŁY