Merck
Aortitis in giant cell arteritis: diagnosis with FDG PET/CT and agreement with CT angiography.
Autoimmunity reviews (2017-09-16)
Mona Hommada, Arsène Mekinian, Pierre-Yves Brillet, Sébastien Abad, Claire Larroche, Robin Dhôte, Olivier Fain, Michael Soussan
ABSTRAKT
To assess the detection rate of aortitis in giant cell arteritis (GCA) with fluorodeoxyglucose positron emission tomography/computed tomography (PET) and to compare the findings with CT angiography (CTA). Fifty-two GCA patients and 27 controls were included. GCA patients had a PET scan at diagnosis (35/52) or during relapse (17/52). Concomitant CTA was performed in 35/52 patients. Aortitis was defined as FDG uptake higher than the liver for PET and wall thickness≥3mm for CTA. Agreement between PET and CTA was evaluated by the kappa coefficient and Spearman correlation coefficient. Aortitis was diagnosed using PET in 40% (14/35) of patients at diagnosis and in 0% of controls (0/27). Agreement was perfect between PET and CT at a patient-based level, and very good at a vascular segment-based level (kappa: 0.72 to 1). PET was positive in 35% (6/17) of patients scanned during GCA relapse, showing aortitis (n=4) and/or articular uptake (n=4). Discrepancies between PET and CT were observed only in relapsing GCA (n=3). Correlation between the maximum standardized uptake value and wall thickness was moderate at diagnosis (r: 0.57 to 0.7) and not statistically significant during relapse. The detection rate of aortitis in GCA patients using PET is 40%, approximately in the range of CTA rates, suggesting that the two techniques have similar sensitivity. PET seems valuable in relapsing GCA, allowing the detection of vascular and articular activities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)