Merck
1, 6-di-O-caffeoyl-β-D-glucopyranoside, a natural compound from Callicarpa nudiflora Hook impairs P2Y
Phytomedicine : international journal of phytotherapy and phytopharmacology (2017-11-22)
Jianjiang Fu, Xiaocui Zhu, Wei Wang, Hong Lu, Zhoumiao Zhang, Ting Liu, Huanjun Xu, Huizheng Fu, Shuangcheng Ma, Yuehua Luo
ABSTRAKT
Platelet activation and subsequent accumulation at sites of vascular injury perform a central role in thrombus formation, which is believed to be the trigger of several cardiovascular diseases, such as atherosclerosis, myocardial infarction and strokes. In this sense, the search for agents that are capable of blocking platelets aggregation has important implications for these diseases. Callicarpa nudiflora (C. nudiflora) Hook is a traditional Chinese medicine herb for eliminating stasis to subdue swelling and hemostasis. Our previous study found several compounds extracted from this herb, including 1, 6-di-O-caffeoyl-β-D-glucopyranoside (CGP), showed inhibitory effects on adenosine diphosphate (ADP) induced platelet aggregation. The aim of current study is confirmation of the anti-platelet effects and elucidation of the probable mechanisms. The experiments were performed on platelet rich plasma freshly isolated from SD rat. ADP, U46619 or arachidonic acid (AA) induced platelet aggregation assay were performed to evaluate the anti-platelet properties of CGP. Activated α CGP inhibited platelet aggregation induced by ADP, U46619 and arachidonic acid (AA), significantly. Furthermore, it is also found that LGP exhibited obvious inhibitory effects on α Collectively, the data presented here demonstrated that CGP, a natural compound from Callicarpa nudiflora Hook, inhibited the development of platelet aggregation and amplification of platelet activation. These inhibitory effects may be associated with its dual-receptor inhibition on P2Y
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate