Merck
Tissue injury of the remnant liver following radiofrequency-assisted partial hepatectomy.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology (2017-11-15)
Petros Ypsilantis, Maria Lambropoulou, Anastasios Karayiannakis, Miroslav Milicevic, Predrag Bulajic, Dimitrios Zacharoulis, Eleni Sioka, Constantinos Simopoulos
ABSTRAKT
To evaluate remnant liver tissue damage in a pig model of radiofrequency (RF)-assisted liver resection employing either the sequential coagulate cut (SCC) Belgrade technique using a monopolar RF electrode or the one using the bipolar Habib-4x device. Sixteen pigs underwent either a) resection of part of the left lateral and left median hepatic lobes employing the SCC (SCC group), the Habib-4X (H group) or the "crushclamp" technique (CC group) or b) sham operation (Sham group). Forty-eight hours later, tissue specimens were excised from the right lateral hepatic lobe for histopathological examination and immunohistochemical assessment of tissue injury, mitosis and inflammation. Histopathologic lesions, apoptotic activity, HSP 40 and TNFα expression were more intense, while mitotic activity was less prominent in the SCC group technique compared to H group. Comparison between CC and H groups suggested the pivotal role of partial hepatectomy (PH) per se in the changes noted in H group. The Habib-4X liver resection technique proved to be less injurious in the remnant liver tissue after PH compared to the SCC technique.