Merck
Synthesis, structure elucidation, and determination of polyhalogenated N-methylpyrroles (PMPs) in blue mussels.
Environmental science and pollution research international (2017-09-25)
Carolin Hauler, Walter Vetter
ABSTRAKT
Polyhalogenated N-methylpyrroles (PMPs) are halogenated natural products (HNPs) recently detected in seagrass, blue mussels, and other marine organisms. In this study, we synthesized 2,3,4,5-tetrachloro-N-methylpyrrole (Cl
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
N-Bromosuccinimide, ReagentPlus®, 99%