Merck
Distinct signaling events promote resistance to mitoxantrone and etoposide in pediatric AML: a Children's Oncology Group report.
Oncotarget (2017-11-23)
Xin Long, Robert B Gerbing, Todd A Alonzo, Michele S Redell
ABSTRAKT
Despite aggressive chemotherapy including mitoxantrone and etoposide, relapse occurs for almost half of children with acute myeloid leukemia (AML). Since both drugs inhibit topoisomerase II and cause DNA double strand breaks, resistance could be achieved by enhanced DNA damage repair (DDR), via homologous recombination (HR) and/or non-homologous end joining (NHEJ). An important source of extrinsic chemoresistance is the bone marrow stroma. We aimed to reveal intrinsic and stroma-induced signaling pathways that contribute to chemoresistance. Sixty diagnostic pediatric AML samples were cultured on or off stromal cells, with or without chemotherapy. We measured apoptosis, DNA damage signaling, and NHEJ/HR pathway activity by FACS analysis of intracellular cleaved PARP, γH2AX, pDNA-PKcs and pATM, respectively. Mitoxantrone strongly increased γH2AX and pDNA-PKcs. Neither chemotherapy drug induced pATM. DNA-PK inhibition alleviated mitoxantrone resistance for AML cells on and off stromal cells. Regarding stroma-induced signaling pathways, ERK1/2 was most consistently activated in primary AML cells by stromal cells. ERK1/2 inactivation partially restored chemosensitivity to AML cells on stromal cells. Additionally, low stroma-induced STAT3 activity and strong stroma-induced mitoxantrone resistance were associated with inferior clinical outcome. Taken together, the NHEJ DDR and ERK1/2 pathways are potential targets for reducing intrinsic and extrinsic chemotherapy resistance in pediatric AML.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Inhibitor DNA-PK V, The DNA-PK Inhibitor V, also referenced under CAS 404009-46-7, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder