Merck
A highly efficient ruthenium catalyst for metathesis reactions.
Angewandte Chemie (International ed. in English) (2002-11-02)
Karol Grela, Syuzanna Harutyunyan, Anna Michrowska
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
nitro-Grela, ≥95%