Merck
Paper-based MoS
Scientific reports (2017-12-14)
Alisha Geldert, Kenry, Chwee Teck Lim
ABSTRAKT
There has been growing interest in the development of paper-based biosensors because their simplicity and low cost are attractive for point-of-care diagnosis, especially in low-resource areas. However, only a limited range of paper materials - primarily chromatography papers - have been incorporated into diagnostics thus far. Here, we investigate the performance of different types of paper in order to develop an aptamer- and MoS