Merck
Favorable Vascular Actions of Angiotensin-(1-7) in Human Obesity.
Hypertension (Dallas, Tex. : 1979) (2017-12-06)
Francesca Schinzari, Manfredi Tesauro, Augusto Veneziani, Nadia Mores, Nicola Di Daniele, Carmine Cardillo
ABSTRAKT
Obese patients have vascular dysfunction related to impaired insulin-stimulated vasodilation and increased endothelin-1-mediated vasoconstriction. In contrast to the harmful vascular actions of angiotensin (Ang) II, the angiotensin-converting enzyme 2 product Ang-(1-7) has shown to exert cardiovascular and metabolic benefits in experimental models through stimulation of the Mas receptor. We, therefore, examined the effects of exogenous Ang-(1-7) on vasodilator tone and endothelin-1-dependent vasoconstriction in obese patients. Intra-arterial infusion of Ang-(1-7) (10 nmol/min) resulted in significant increase in unstimulated forearm flow (
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
BQ-123, ≥99%, sodium salt, lyophilized powder