Merck
Peptide nucleic acid as a selective recognition element for electrochemical determination of Hg
Bioelectrochemistry (Amsterdam, Netherlands) (2017-10-22)
Agnieszka Bala, Łukasz Górski
ABSTRAKT
A novel electrochemical PNA-based biosensor for the determination of Hg
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hexaammineruthenium(III) chloride, 98%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM