Merck
Kinase-independent role of nuclear RIPK1 in regulating parthanatos through physical interaction with PARP1 upon oxidative stress.
Biochimica et biophysica acta (2017-10-11)
Ki-Hong Jang, Taeik Jang, Eunji Son, Soonjin Choi, Eunhee Kim
ABSTRAKT
Regulated necrosis occurs in various pathophysiological conditions under oxidative stress. Here, we report that receptor-interacting protein kinase 1 (RIPK1), a key player in one type of regulated necrosis (necroptosis), also participates in another type of poly (ADP-ribose) polymerase 1 (PARP1)-dependent regulated necrosis (parthanatos). Various biological signatures of parthanatos were significantly attenuated in Ripk1
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)