Merck
Abiotic synthesis of purine and pyrimidine ribonucleosides in aqueous microdroplets.
Proceedings of the National Academy of Sciences of the United States of America (2017-12-20)
Inho Nam, Hong Gil Nam, Richard N Zare
ABSTRAKT
Aqueous microdroplets (<1.3 µm in diameter on average) containing 15 mM d-ribose, 15 mM phosphoric acid, and 5 mM of a nucleobase (uracil, adenine, cytosine, or hypoxanthine) are electrosprayed from a capillary at +5 kV into a mass spectrometer at room temperature and 1 atm pressure with 3 mM divalent magnesium ion (Mg
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Inosine, ≥99% (HPLC)